MacConkey Agar, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C6132
MacConkey Agar, CRITERION™, Hardy Diagnostics
Catalog # CA89405-632
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ MacConkey Agar is recommended for use as a selective and differential medium for the isolation of gram-negative bacilli (including coliform organisms and enteric pathogens), on the basis of lactose fermentation
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bucket
- Taille:2 kg
- Cat. No.:CA89405-632
- Storage temperature:–2-30°C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



