Lactose Broth, CRITERION™, Hardy Diagnostics
122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®
Catalog # CA89405-528
Supplier: HARDY DIAGNOSTICS CA
: C5940
Lactose Broth, CRITERION™, Hardy Diagnostics
Catalog # CA89405-528
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Lactose Broth is used to detect coliforms in food, dairy products, and water, and to preenrich samples for the recovery of Salmonella species
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Zip-pouch to make 2 L, Mylar®
- Taille:122.4 g
- Final Volume:2 L
- Cat. No.:CA89405-528
- Storage temperature:–2-30°C



