Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Extract Broth, CRITERION™, Hardy Diagnostics | Avantor

Extract Broth, CRITERION™, Hardy Diagnostics

2 kg, Bucket
Catalog # CA89405-444
Extract Broth, CRITERION™, Hardy Diagnostics
Catalog # CA89405-444
Supplier:  
CAS Number:  
Extract Broth, CRITERION™, Hardy Diagnostics
Extract Broth, CRITERION™, Hardy Diagnostics

2 kg, Bucket

CAS Number:  
Discontinued
Product Details & Documents

CRITERION™ Extract Broth for the cultivation of microorganisms.

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

Dehydratedculture media with engineered container.

 : BSE Free

Spécifications

Spécifications

  • Type de conditionnement:
    Bucket
  • Taille:
    2 kg
  • Cat. No.:
    CA89405-444
  • Storage temperature:
    –2-30°C

Spécifications