EC Medium, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: C5692
EC Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-424
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ EC Medium is recommended for the detection of coliforms at 35 degrees C
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
and Escherichia coli and fecal coliforms at 45.5 degrees C. Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bucket
- Taille:2 kg
- Cat. No.:CA89405-424
- Storage temperature:–2-30°C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



