Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
EC Medium, CRITERION™, Hardy Diagnostics | Avantor

EC Medium, CRITERION™, Hardy Diagnostics

500 g, Bottle, wide mouth
Catalog # CA89405-422
Fournisseur #:  C5691
EC Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-422
CAS Number:  
EC Medium, CRITERION™, Hardy Diagnostics
EC Medium, CRITERION™, Hardy Diagnostics

500 g, Bottle, wide mouth

CAS Number:  
Price
$381.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

CRITERION™ EC Medium is recommended for the detection of coliforms at 35 degrees C

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

and Escherichia coli and fecal coliforms at 45.5 degrees C. Dehydratedculture media with engineered container.

Certifications: BSE Free

Spécifications

Spécifications

  • Type de conditionnement:
    Bottle, wide mouth
  • Taille:
    500 g
  • Cat. No.:
    CA89405-422
  • Storage temperature:
    –2-30°C

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.