Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics

Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics

2 kg, Bucket
Catalog # CA89405-366
Supplier #:  C5572
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-366
CAS Number:  
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics

2 kg, Bucket

CAS Number:  
Price
$3,680.00
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents

CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications

Dehydratedculture media with engineered container.

Certifications: BSE Free

Spécifications

Spécifications

  • Type de conditionnement:
    Bucket
  • Taille:
    2 kg
  • Cat. No.:
    CA89405-366
  • Storage temperature:
    –2-30°C

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.