Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
10 kg, Bucket
Catalog # CA89405-360
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: C5563
Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
Catalog # CA89405-360
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Deoxycholate Lactose Agar for cultivation, maintenance, fermentation reaction and motility determination of bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bucket
- Taille:10 kg
- Cat. No.:CA89405-360
- Storage temperature:–2-30°C
Spécifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



