Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
: C5561
CRITERION™ Deoxycholate Lactose Agar for cultivation, maintenance, fermentation reaction and motility determination of bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
- Type de conditionnement:Bottle, wide mouth
- Taille:500 g
- Cat. No.:CA89405-356
- Storage temperature:–2-30°C