Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
  CA89405-356
 :  C5561
Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
  CA89405-356
 :  
Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
 :  
$259.00
Each
CRITERION™ Deoxycholate Lactose Agar for cultivation, maintenance, fermentation reaction and motility determination of bacteria
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
 : BSE Free
  • Type de conditionnement:
    Bottle, wide mouth
  • Taille:
    500 g
  • Cat. No.:
    CA89405-356
  • Storage temperature:
    –2-30°C