Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Fluid D, USP

Fluid D, USP

1 L, PC bottle, narrow mouth, screw top, –2...30 °C
Catalog # CA52428-862
Supplier #:  U115
Fluid D, USP
Catalog # CA52428-862
CAS Number:  
Fluid D, USP
Fluid D, USP

1 L, PC bottle, narrow mouth, screw top, –2...30 °C

CAS Number:  
Price
$467.00
Log in pour la disponibilité
Pack of 10
Product Details & Documents

Fluid D is a rinsing and diluting fluid for use in USP testing protocols.

  • Rinsing and diluting fluid
  • Contains peptic digest of animal tissue and polysorbate 80

A rinsing and diluting fluid for use in USP testing protocols. pH 7.1. Contains peptic digest of animal tissue and polysorbate 80.

Spécifications

Spécifications

  • Type de conditionnement:
    PC bottle, narrow mouth, screw top
  • Taille:
    1 L
  • Température de stockage:
    –2...30 °C

Spécifications