Copper(II) Nitrate Trihydrate
Copper(II) Nitrate Trihydrate, Reagent, 100 g
Supplier: Ward's Science
Fournisseur #: CC0515-100GR
Jump to:
Product Details & Documents
CAS Number: 10031-43-3
Formula Weight: 241.60
Formula: Cu(NO3)2·3H2O
Hazard Info: Toxic, Oxidizer
Density (g/mL): 2.32
Freezing Point (°C): 114.5
Solubility: Water and Alcohol
Synonyms: Copper Nitrate, Trihydrate, Copper(II) Nitrate 3-Hydrte
Shelf Life (months): 12
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Copper(II) Nitrate Trihydrate
- Taille:100 g
- Chemical/Physical Specifications:Crystal
- Grade:Reagent



