Simulaids® CPR Torso | Avantor
Simulaids® CPR Torso
Replacement Mouth Nose Pieces ( Pk/10)
Catalog # 470103-408
Supplier:   Nasco
Fournisseur #:  100-2023
Simulaids® CPR Torso
Catalog # 470103-408
Supplier:  Nasco
Simulaids® CPR Torso
Simulaids® CPR Torso
Replacement Mouth Nose Pieces ( Pk/10)
Price
$206.35
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Lifelike anatomical landmarks such as a carotid pulse, sternum, ribcage, and sub sternal notch provide realistic CPR practice.
  • Lightweight, easy-to-transport torso
  • Realistic anatomical landmarks
  • Easy to use
  • Unit includes a carrying case
The torso features an airway ball and valve, which allows the lungs to inflate only if the head is extended and accurately positioned, so students learn and practice the proper procedure. The lightweight model is easy to transport and includes a soft carrying case with kneeling pads and simple-to-replace mouth/nose pieces.
The version with electronics has all the same features and also includes a light controller that confirms correct hand placement, ventilation, and compression.
Spécifications
  • Désignation:
    Replacement Mouth Nose Pieces ( Pk/10)
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Mouth Nose Pieces ( Pk/10)
    Replacement Mouth Nose Pieces ( Pk/10)
    470103-408
    Log in pour la disponibilité
    Pack of 10 - $206.35