Wright-Giemsa stain
1 L (33.8 oz.)
Catalog # CA84000-182
Supplier: Thermo Scientific
Fournisseur #: 89013
Jump to:
Product Details & Documents
Rapidly and conveniently stain blood smears with Epredia™ Wright Giemsa Stain.
Synonymes: Wright-Giemsa Solution
- Numéro MDL:MFCD00132593
Spécifications
Spécifications
- Taille:1 L (33.8 oz.)
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



