Dehydrant Blends, Epredia
Dehydrant 80%, Poly Bottle, 4 x 1 gal.
CA84000-050
: 6401
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
- Blend of ethyl, methyl and isopropyl alcohols
- Can be used as a dye or stain solvent
- Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.
- Type de conditionnement:Poly Bottle
- Désignation:Dehydrant 80%
- Taille:4 x 1 gal.