Dehydrant Blends, Epredia
Dehydrant 95%, Poly Bottle, 1 gal.
Catalog # CA84000-046
Supplier: Thermo Scientific
: 6301
Jump to:
Product Details & Documents
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
- Blend of ethyl, methyl and isopropyl alcohols
- Can be used as a dye or stain solvent
- Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.
Spécifications
Spécifications
- Type de conditionnement:Poly Bottle
- Désignation:Dehydrant 95%
- Taille:1 gal.