Dehydrant Blends, Epredia
Dehydrant 95%, Poly Bottle, 1 gal.
Catalog # CA84000-046
Supplier:   Thermo Scientific
 :  6301
Dehydrant Blends, Epredia
Catalog # CA84000-046
Supplier:  Thermo Scientific
CAS Number:  
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia
Dehydrant 95%, Poly Bottle, 1 gal.
CAS Number:  
Price
$502.33
Log in pour la disponibilité
Case of 4 (0.25 .US Gal)
Product Details & Documents
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.
Spécifications
Spécifications
  • Type de conditionnement:
    Poly Bottle
  • Désignation:
    Dehydrant 95%
  • Taille:
    1 gal.
Spécifications