Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
Product Details & Documents
Eppendorf® Reference® 2 multi-channel pipettes feature an innovative single-button operation, offering users precise, efficient, and ergonomic liquid handling that ensures reproducible results. Easy decontamination and a long service life will make them indispensable for high-throughput applications.
- Operated with just one button for volume selection, pipetting, and tip ejection
- 4-digit display with magnifying window for quick and secure volume setting and identification (clearly visible from every angle), incl. volume lock
- The unique smooth surface and full autoclavability guarantee efficient decontamination - making it ideal when working under sterile conditions
- Temporary adjustment allows for an easy adjustment to accurately pipette challenging liquids or in changing external conditions without the need for a full calibration
- Spring-loaded tip cones, which can be switched on/off, improve user-to-user reproducibility and ergonomics by lowering the tip attachment and ejection force
Pipetting type: air-cushion system
Operating type: mechnical
Compatible tips: epT.I.P.S.®, Universal tips
Applications include forward pipetting, reverse pipetting, removal of supernatants, sample mixing, phase extraction, and filling of plates, gels, and reaction vessels.
Ordering information: All Reference® 2 pipettors are supplied with an ep.T.I.P.S. box.
Spécifications
- Volume:10 - 100 µl
- Désignation:Reference® 2, 12-channel, variable volume, spring-loaded tip
- Increments:0.1 µl
- No. of Channels:12
- Color Code:Yellow
- Cat. No.:CA10032-660
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Informations sur le produit
- Catalog #CapacityAvailabilityPrice
- Reagent reservoir tip-tub
Informations sur le produit
- Catalog #AvailabilityPrice
- Pipette carousel 2
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



