Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
Reference® 2, 12-channel, variable volume, spring-loaded tip, 0.5 - 10 µl, 0.01 µl, Medium gray, 12
Supplier: Eppendorf
: 4926000050
Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
Catalog # CA10032-658
Supplier: Eppendorf
Jump to:
Product Details & Documents
Eppendorf® Reference® 2 multi-channel pipettes feature an innovative single-button operation, offering users precise, efficient, and ergonomic liquid handling that ensures reproducible results. Easy decontamination and a long service life will make them indispensable for high-throughput applications.
- Operated with just one button for volume selection, pipetting, and tip ejection
- 4-digit display with magnifying window for quick and secure volume setting and identification (clearly visible from every angle), incl. volume lock
- The unique smooth surface and full autoclavability guarantee efficient decontamination - making it ideal when working under sterile conditions
- Temporary adjustment allows for an easy adjustment to accurately pipette challenging liquids or in changing external conditions without the need for a full calibration
- Spring-loaded tip cones, which can be switched on/off, improve user-to-user reproducibility and ergonomics by lowering the tip attachment and ejection force
Pipetting type: air-cushion system
Operating type: mechnical
Compatible tips: epT.I.P.S.®, Universal tips
Applications include forward pipetting, reverse pipetting, removal of supernatants, sample mixing, phase extraction, and filling of plates, gels, and reaction vessels.
: All Reference® 2 pipettors are supplied with an ep.T.I.P.S. box.
Specifications
- Volume:0.5 - 10 µl
- Désignation:Reference® 2, 12-channel, variable volume, spring-loaded tip
- Increments:0.01 µl
- No. of Channels:12
- Color Code:Medium gray
Accessories
Product Information
- Catalog #CapacityAvailabilityPrice
- Reagent reservoir tip-tub
Product Information
- Catalog #AvailabilityPrice
- Pipette carousel 2