WHEATON® Screw Thread Vials, DWK Life Sciences
Tubular Vials, 10.00 ml, 22×55 mm, Borosilicate glass, Clear, Non, 20-400
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: W219366
WHEATON® Screw Thread Vials, DWK Life Sciences
Catalog # 10070-716
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
The tubular design provides excellent clarity and dimensional consistency from vial to vial.
- Ideal for lyophilization
- Alternative to serum bottles and vials
- Heavy walled glass provides impact resistance
- Screw thread design eliminates need for crimping tools
These screw thread vials fits with I-Loc™ closure or screw cap with thin flange stopper.
Specifications
- Avec bouchon:Non
- Taille du bouchon:20-400
- Capacité:10.00 ml
- Color:Clear
- Désignation:Tubular Vials
- Matière:Borosilicate glass
- Diameter ×H:22×55 mm



