TempDex 710, Nitrile Gloves, Mapa Professional
11, Yellow, 26 cm (10³⁄₁₆")
Catalog # 10067-644
Jump to:
Discontinued
Product Details & Documents
These high dexterity and thermal protection gloves are suitable for handling of composite materials after heat processing in ceramics, plastics and composite industries.
- Nitrile coated palm and dotted embossing for better insulation and grip
- Thin, anatomically-shaped glove ensures dexterity
- Long lasting and abrasion resistant
Ideal for use in ceramics, plastics, composite and construction industries.
Spécifications
- Glove Color:Yellow
- Taille du gant:11
- Longueur:26 cm (10³⁄₁₆")