TempDex 710, Nitrile Gloves, Mapa Professional | Avantor
TempDex 710, Nitrile Gloves, Mapa Professional
7, Yellow, 26 cm (10³⁄₁₆")
Catalog # 10067-640
Supplier:   MAPA
Fournisseur #:  34710037
TempDex 710, Nitrile Gloves, Mapa Professional
Catalog # 10067-640
Supplier:  MAPA
TempDex 710, Nitrile Gloves, Mapa Professional
TempDex 710, Nitrile Gloves, Mapa Professional
7, Yellow, 26 cm (10³⁄₁₆")
Price
$19.19
Log in pour la disponibilité
Each Case of 50
Product Details & Documents
These high dexterity and thermal protection gloves are suitable for handling of composite materials after heat processing in ceramics, plastics and composite industries.
  • Nitrile coated palm and dotted embossing for better insulation and grip
  • Thin, anatomically-shaped glove ensures dexterity
  • Long lasting and abrasion resistant
Ideal for use in ceramics, plastics, composite and construction industries.
Spécifications
  • Glove Color:
    Yellow
  • Taille du gant:
    7
  • Longueur:
    26 cm (10³⁄₁₆")