CF® Dye BCN, Biotium
CF® Dye BCN, Biotium
CF®488A BCN, 490/515 nm, Impermeant, 0.5 mg
Catalog # 10066-546
Supplier:   Biotium
Supplier #:  92075
CF® Dye BCN, Biotium
Catalog # 10066-546
Supplier:  Biotium
CAS Number:  
CF® Dye BCN, Biotium
CF® Dye BCN, Biotium
CF®488A BCN, 490/515 nm, Impermeant, 0.5 mg
CAS Number:  
Price
$699.79
Log in pour la disponibilité
Each
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
CF® Dyes with a BCN group react with azides via a copper-free bioorthogonal reaction to label target molecules for surface staining of live cells or cell extracts. Membrane-permeant CF® Dye BCN conjugates for intracellular labeling reactions are also available.
  • Fluorescent labeling of azide groups on target molecules via copper-free bioorthonal chemistry
  • Alternative to fluorescently labeled DIBO and DBCO
  • Suitable for surface or intracellular labeling of live cells or cell extracts
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labeling
  • Available in several CF® Dye options including membrane-permeant forms for intracellular labeling
CF® Dyes BCN (bicyclo[6.1.0]nonyne) derivatives are alternatives to fluorescently labeled DIBO and DBCO. They react with azides to form 1,2,3-triazole by copper-free bioorthogonal 1,3-dipolar Huisgen cycloaddition. Useful for fluorescent surface labeling of live cells or cell extracts, particularly when there are concerns about native protein function loss with copper. Membrane-permeant CF® Dye BCN options are also available for intracellular labeling reactions.
Available CF® Dye BCN derivatives: CF®405S, CF®405M, CF®488A, CF®568, CF®594, CF®640R, CF®647, CF®680, and CF®680R including membrane-permeant CF®440 BCN, CF®500 BCN, and CF®650 BCN. Biotium’s next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Spécifications
Spécifications
  • Désignation:
    CF®488A BCN
  • Taille:
    0.5 mg
  • Ex/Em:
    490/515 nm
  • Membrane:
    Impermeant