Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Barium Nitrate

Barium Nitrate

Barium Nitrate, Reagent, 500 g
  470300-348
Supplier #:  BB0055-500GR
Barium Nitrate
  470300-348
 :  
Barium Nitrate
Barium Nitrate

Barium Nitrate, Reagent, 500 g

 :  
 
$174.10
   
Each (500g)
Minimum order quantity is 1
 

CAS Number: 10022-31-8
Formula Weight: 261.35
Formula: Ba(NO3)2
Hazard Info: Oxidizer, Toxic
Density (g/mL): 3.24
Boiling Point (°C): Decomposes
Freezing Point (°C): 592
Solubility: Water
Synonyms: Barium Dinitrate, Nitric Acid Barium Salt
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 

 

  • Désignation:
    Barium Nitrate
  • Taille:
    500 g
  • Chemical/Physical Specifications:
    Crystal
  • Grade:
    Reagent

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.