Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Ammonium Nitrate | Avantor

Ammonium Nitrate

Ammonium Nitrate, Reagent, 100 g
Catalog # 470300-234
Supplier:   Ward's Science
Fournisseur #:  AA0295-100GR
Ammonium Nitrate
Catalog # 470300-234
Supplier:  Ward's Science
CAS Number:  
Ammonium Nitrate
Ammonium Nitrate

Ammonium Nitrate, Reagent, 100 g

CAS Number:  
Price
$28.30
Log in pour la disponibilité
Each (100g)
La quantité minimale de commande est de 1
Product Details & Documents

CAS Number: 6484-52-2
Formula: NH4NO3
Density: 1.73 g/mL
Boiling and Freezing Point: Decomposes, 169 °C
Solubility: Water, Alcohol, and Alkalies
Synonyms: Nitric Acid Ammonium Salt
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: The product is a toxic, reactive irritant.

Spécifications

Spécifications

  • Désignation:
    Ammonium Nitrate
  • Taille:
    100 g
  • Chemical/Physical Specifications:
    Granular
  • Grade:
    Reagent

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.