Ammonium Nitrate
Le produit 470300-234 est similaire à 470300-232 mais le fabricant et les spécifications peuvent varier.
Product Details & Documents
CAS Number: 6484-52-2
Formula: NH4NO3
Density: 1.73 g/mL
Boiling and Freezing Point: Decomposes, 169 °C
Solubility: Water, Alcohol, and Alkalies
Synonyms: Nitric Acid Ammonium Salt
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: The product is a toxic, reactive irritant.
Spécifications
Spécifications
- Désignation:Ammonium Nitrate
- Taille:2.5 kg
- Chemical/Physical Specifications:Granular
- Grade:Laboratory
Spécifications
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



