Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sodium Nitrate | Avantor

Sodium Nitrate

Sodium Nitrate, Laboratory, 500 g
Catalog # 470302-640
Supplier:   Ward's Science
Fournisseur #:  SS0690-500G
Sodium Nitrate
Catalog # 470302-640
Supplier:  Ward's Science
CAS Number:  
Sodium Nitrate
Sodium Nitrate

Sodium Nitrate, Laboratory, 500 g

CAS Number:  
Price
$29.80
Log in pour la disponibilité
Each (500g)
 
Product Details & Documents

CAS Number: 7631-99-4
Formula Weight: 84.99
Formula: NaNO3
Hazard Info: Oxidizer, Explosive, Toxic
Density (g/mL): 2.261
Freezing Point (°C): 307
Solubility: Water and Alcohol
Synonyms: Nitrate of Soda, Chile Saltpeter
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Sodium Nitrate
  • Taille:
    500 g
  • Chemical/Physical Specifications:
    Crystal
  • Grade:
    Laboratory

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.