Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sodium Hydroxide | Avantor

Sodium Hydroxide

Sodium Hydroxide, Reagent, 100 g
Catalog # 470302-554
Supplier:   Ward's Science
Fournisseur #:  SS0550-100GR
Sodium Hydroxide
Catalog # 470302-554
Supplier:  Ward's Science
CAS Number:  
Sodium Hydroxide
Sodium Hydroxide

Sodium Hydroxide, Reagent, 100 g

CAS Number:  
Price
$19.60
Log in pour la disponibilité
Each (100g)
La quantité minimale de commande est de 1
Product Details & Documents

CAS Number: 1310-73-2
Formula: NaOH
Density: 2.13 g/mL
Boiling and Freezing Point: 1390°C, 318°C
Solubility: Water and Alcohol
Synonyms: Caustic Soda
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is corrosive and toxic.

Spécifications

Spécifications

  • Désignation:
    Sodium Hydroxide
  • Taille:
    100 g
  • Chemical/Physical Specifications:
    Pellet
  • Grade:
    Reagent

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.