Polypropylene Beaker Sets
50 / 100 / 250 / 500 / 1000 ml, Set of 5, 5 / 10 / 25 / 50 / 100 ml
Catalog # 490012-214
Supplier #: 522135-0001
Jump to:
Discontinued
Product Details & Documents
Unbreakable Polypropylene
- Autoclavable
- Chemical-resistant
These beaker sets are autoclavable up to 145°C and chemical resistant. All feature pour spouts and metric graduation scales.
Specifications
- Intervalle de graduation:5 / 10 / 25 / 50 / 100 ml
- Capacité:50 / 100 / 250 / 500 / 1000 ml
- Désignation:Set of 5