Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Grey
490010-386
: T312-9
Color coded inserts fit precisely into the cap of the Cryovial® tubes for color identification.
Made of polypropylene
: T312-7 contains 100 pieces each of white, blue, red, green and yellow inserts packaged in individual bags.
- Color:Grey