Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Assorted
  490010-382
 :  T312-7
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
  490010-382
 :  
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Assorted
Color coded inserts fit precisely into the cap of the Cryovial® tubes for color identification.
Made of polypropylene
 : T312-7 contains 100 pieces each of white, blue, red, green and yellow inserts packaged in individual bags.
  • Color:
    Assorted