Cryogenic Printer Labels, Brady
Cryogenic Printer Labels, Brady
Cryogenic printer labels, Nylon Cloth, Black on white, 48.3×25.4 mm (1.9×1")
Catalog # 490006-452
Cryogenic Printer Labels, Brady
Catalog # 490006-452
Supplier:  
Cryogenic Printer Labels, Brady
Cryogenic Printer Labels, Brady
Cryogenic printer labels, Nylon Cloth, Black on white, 48.3×25.4 mm (1.9×1")
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
LABXPERT LABELS B-499 MATERIAL Catalog #CA80090-400
Product CA80090-400 is a replacement for 490006-452 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Cryogenic labels for tubes, vials, and canes.
Chemical and stain resistant labels for slides and cassettes:
Temperatures from –80 to 121 °C (–112 to 250 °F)
B-488 Slide label with moderate chemical resistance
B-498 Tissue cassette label for re-identification of processed paraffin blocks.
General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more:
B-499 For cryo boxes, freezer racks, petri dishes, bottles, flasks, microplates, and bags, working temperatures from –196 to 121 °C (–321 to 250 °F)
B-595 General purpose label for drawers, shelves, and floor.
Spécifications
  • Color:
    Black on white
  • Désignation:
    Cryogenic printer labels
  • Dimensions:
    48.3×25.4 mm (1.9×1")
  • Matière:
    Nylon Cloth