Lithium Chloride
Lithium Chloride, Laboratory, 100 g
Supplier: Ward's Science
Fournisseur #: LL0220-100G
Jump to:
Product Details & Documents
CAS Number: 7447-41-8
Formula: LiCl
Density: 2.068 g/mL
Boiling and Freezing Point: 1382 °C, 614 °C
Solubility: Water and Alcohol
Shelf Life: 12 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Lithium Chloride
- Taille:100 g
- Chemical/Physical Specifications:Granular
- Grade:Laboratory
Spécifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



