Microscope Lens Solution | Avantor
Microscope Lens Solution
Microscope Lens Solution, 100 ml
Catalog # 470301-371
Supplier:   Ward's Science
Fournisseur #:  MM0565-100ML
Microscope Lens Solution
Catalog # 470301-371
Supplier:  Ward's Science
Microscope Lens Solution
Microscope Lens Solution
Microscope Lens Solution, 100 ml
Price
$19.20
Log in pour la disponibilité
Each (100ml)
La quantité minimale de commande est de 1
Product Details & Documents
Shelf Life (months): 36
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
  • Désignation:
    Microscope Lens Solution
  • Taille:
    100 ml