Rtx®-5 GC Columns, Restek
Rtx-5, Capillaire, 0.50 µm, 0.32 mm, 10 m
Catalog # 10059-926
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
A low-polarity general-purpose column for the analysis of pesticides, PCBs, aromatic hydrocarbons, drugs, and essential oils.
- Extremely low bleed
- Low-polarity column
- Internal dimensions and film thickness vary between models
Equivalent to USP G27, G36 phases. All residual catalysts and low molecular weight fragments are removed from the Rtx®-5 polymer, providing a tight mono-modal distribution and extremely low bleed. Phase composition: Crossbond® 5% diphenyl, 95% dimethylpolysiloxane.
Specifications
- Type de colonne:Capillaire
- Épaisseur du film:0.50 µm
- Ø int.:0.32 mm
- Désignation:Rtx-5
- Longueur:10 m
- Cat. No.:10059-926
- Temperature limit:–60...350 °C



