Standard For Osmometer, Advanced Instruments
Standard For Osmometer, Advanced Instruments
1000 mOsm Calibration Standard, 10×2 ml
Catalog # CA89369-186
Supplier #:  3MA100
Standard For Osmometer, Advanced Instruments
Catalog # CA89369-186
CAS Number:  
Standard For Osmometer, Advanced Instruments
1000 mOsm Calibration Standard, 10×2 ml
CAS Number:  
Price
$402.56
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Calibration standard is used to check the perforance of osmometer and test methods.
Documents
Specifications
Specifications
  • Désignation:
    1000 mOsm Calibration Standard
  • Taille:
    10×2 ml