Corning® Spheroid Microplates, Corning
Spheroid Microplates, 96, 75 - 300 µl, Round bottom, Black/Clear, Sterile, Ultra-low attachment
: Corning
Supplier #: 4520
Corning® Spheroid Microplates, Corning
10037-558
: Corning
Product available on GSA Advantage®
PS, black opaque microplate with optically clear bottom, with lid. The Ultra-Low Attachment surface is a covalently bound hydrogel layer that is hydrophilic and neutrally charged. The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes embryoid body formation from ES cells.
- Well geometry aids formation of spheroids in centre of well
- Non cytotoxic, biologically inert and non degradable surface
- Sterilized by gamma radiation
- Nb de puits:96
- Sterility:Sterile
- Color:Black/Clear
- Désignation:Spheroid Microplates
- Working Volume:75 - 300 µl
- Surface Treatment:Ultra-low attachment
- Bottom Style:Round bottom
- Cat. No.:10037-558
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



