Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes | Avantor
Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
Reference® 2, 12-channel, variable volume, spring-loaded tip, 0.5 - 10 µl, 0.01 µl, Light orange, 12
Catalog # 10032-658
   Low Manufacturing Impact
Fournisseur #:  4926000026
Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
Catalog # 10032-658
Supplier:  
   Low Manufacturing Impact
Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
Eppendorf® Reference® 2 Mechanical Multi-Channel Pipettes
Reference® 2, 12-channel, variable volume, spring-loaded tip, 0.5 - 10 µl, 0.01 µl, Light orange, 12
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VWR PIPETTE EHP 12CHNL 1-10UL Catalog #89079-952
Le produit 89079-952 est similaire à 10032-658 mais peut avoir un fabricant et/ou un emballage différents.
Product Details & Documents
Eppendorf® Reference® 2 multi-channel pipettes feature an innovative single-button operation, offering users precise, efficient, and ergonomic liquid handling that ensures reproducible results. Easy decontamination and a long service life will make them indispensable for high-throughput applications.
  • Operated with just one button for volume selection, pipetting, and tip ejection
  • 4-digit display with magnifying window for quick and secure volume setting and identification (clearly visible from every angle), incl. volume lock
  • The unique smooth surface and full autoclavability guarantee efficient decontamination - making it ideal when working under sterile conditions
  • Temporary adjustment allows for an easy adjustment to accurately pipette challenging liquids or in changing external conditions without the need for a full calibration
  • Spring-loaded tip cones, which can be switched on/off, improve user-to-user reproducibility and ergonomics by lowering the tip attachment and ejection force
Pipetting type: air-cushion system
Operating type: mechnical
Compatible tips: epT.I.P.S.®, Universal tips
Applications include forward pipetting, reverse pipetting, removal of supernatants, sample mixing, phase extraction, and filling of plates, gels, and reaction vessels.
Ordering information: All Reference® 2 pipettors are supplied with an ep.T.I.P.S. box.
Spécifications
  • Volume:
    0.5 - 10 µl
  • Désignation:
    Reference® 2, 12-channel, variable volume, spring-loaded tip
  • Increments:
    0.01 µl
  • No. of Channels:
    12
  • Color Code:
    Light orange
  • Cat. No.:
    10032-658
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Informations sur le produit
  • Catalog #
    Capacity
    Availability
    Price
  • Reagent reservoir tip-tub
    Reagent reservoir tip-tub
    CA53513-048
    50 - 60 ml
    Log in pour la disponibilité
    Pack of 10 - $108.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Pipette carousel 2
    Pipette carousel 2
    76204-774
    Log in pour la disponibilité
    Each - $365.00
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.