VWR® Generator Probes and Replacement Components
VWR® Saw-Tooth Generator Probe, 7×95 mm, For 1.5 - 10 ml Tubes, Model 200; 250; 15D; 25D
Catalog # 10032-566
Supplier: VWR International
VWR® Generator Probes and Replacement Components
Catalog # 10032-566
Supplier: VWR International
Jump to:
Product Details & Documents
Spécifications
- Désignation:VWR® Saw-Tooth Generator Probe, 7×95 mm
- For Use With:For 1.5 - 10 ml Tubes, Model 200; 250; 15D; 25D
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



