MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: Just dip into sample, add reagent, wait and measure visually against colour scale
- Immediate results within 2 minutes enables fast decisions
- Portable: Can be used inside or outside the laboratory
The phosphate test is suitable for determining orthophosphate in wastewater, soil samples, fertilizers and food.
Gradation: 10 - 25 - 50 - 100 - 250 - 500 mg/l PO₄³⁻
- Type de conditionnement:Plastic Box(es)
- Désignation:Phophate test strips
- Taille:1 pc
- Range:10 - 500 mg/L PO₄³⁻
- No. of Tests:100