Ohaus® Starter Electrodes
Ohaus® Starter Electrodes
STCON3: 4-Ring conductivity electrode, Series 300C, 3100C
  470020-480
 :   Ohaus
Supplier #:  83033972
Ohaus® Starter Electrodes
  470020-480
 :  Ohaus
Ohaus® Starter Electrodes
Ohaus® Starter Electrodes
STCON3: 4-Ring conductivity electrode, Series 300C, 3100C
$614.20
Each
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes.
  • Can be used in conjunction with Ohaus® Starter Series
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes that can be used in conjunction with Ohaus® Starter Series 2100 and 3100 bench and Ohaus® Starter 300 Series Portable meters.
Ordering information: Ohaus® 2100 & 3100 Starter Bench Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 3100C Starter Bench Meter Compatible Electrodes are STCON3. Ohaus® 300 Starter Portable Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 300C Starter Portable Meter Compatible Electrodes are STCON3. Ohaus® 300D Starter Portable Meter Compatible Electrodes are STTEMP30, STD011.
  • Désignation:
    STCON3: 4-Ring conductivity electrode
  • For Use With:
    Series 300C, 3100C
Accessories
  •  
    Description
    For Use With
  • 77679-490
    Maintenance kit
    STDO11: DO electrode
    Each - $186.24