Ohaus® Starter Electrodes | Avantor
Ohaus® Starter Electrodes
STTEMP30: Temperature electrode, Series 300, 300D, 2100, 3100
Catalog # 470020-416
Fournisseur #:  83033970
Ohaus® Starter Electrodes
Catalog # 470020-416
Supplier:  
Ohaus® Starter Electrodes
Ohaus® Starter Electrodes
STTEMP30: Temperature electrode, Series 300, 300D, 2100, 3100
Discontinued
Product Details & Documents
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes.
  • Can be used in conjunction with Ohaus® Starter Series
The Ohaus® Starter Series of electrodes include pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO) and temperature electrodes that can be used in conjunction with Ohaus® Starter Series 2100 and 3100 bench and Ohaus® Starter 300 Series Portable meters.
 : Ohaus® 2100 & 3100 Starter Bench Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 3100C Starter Bench Meter Compatible Electrodes are STCON3. Ohaus® 300 Starter Portable Meter Compatible Electrodes are ST210, ST320, STTEMP30. Ohaus® 300C Starter Portable Meter Compatible Electrodes are STCON3. Ohaus® 300D Starter Portable Meter Compatible Electrodes are STTEMP30, STD011.
Spécifications
  • Désignation:
    STTEMP30: Temperature electrode
  • For Use With:
    Series 300, 300D, 2100, 3100
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Maintenance kit
    Maintenance kit
    77679-490
    STDO11: DO electrode
    Log in pour la disponibilité
    Each - 186,24 $