Mini Benchtop Coolers
–12 °C Mini Cooler with clear lid, Blue, 225×165×110 mm, 96
Catalog # 89511-800
Supplier: Heathrow Scientific
Fournisseur #: HS120076
Jump to:
Product Details & Documents
Ideal for keeping samples, PCR reagents, enzymes, and other biochemical reagents cool while working at the bench.
- Protects temperature-sensitive samples at the bench or in the freezer
- Polycarbonate construction with rubber non-slip feet
- Gridded and keyed one-position lids for easy sample identification
- Temperature-retention graph included
Filled with insulating gel, the Mini Coolers can also be used for controlled freezing of samples with longer temperature retention.
Spécifications
- Color:Blue
- Désignation:–12 °C Mini Cooler with clear lid
- Dimensions:225×165×110 mm
- No. of Places:96
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



