Mini Benchtop Coolers
0 °C Mini Cooler with gel filled lid, Green, 245×105×100 mm, 32
Catalog # 89511-792
Supplier: Heathrow Scientific
: HS120072
Jump to:
Product Details & Documents
Ideal for keeping samples, PCR reagents, enzymes, and other biochemical reagents cool while working at the bench.
- Protects temperature-sensitive samples at the bench or in the freezer
- Polycarbonate construction with rubber non-slip feet
- Gridded and keyed one-position lids for easy sample identification
- Temperature-retention graph included
Filled with insulating gel, the Mini Coolers can also be used for controlled freezing of samples with longer temperature retention.
Spécifications
- Color:Green
- Désignation:0 °C Mini Cooler with gel filled lid
- Dimensions:245×105×100 mm
- No. of Places:32



