Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific | Avantor

Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific

Blocking Buffer, 100 mL, ELISA, Western Blot, Dot Blot, Immunohistochemistry
Catalog # CAPI37578
Supplier:   Thermo Scientific
Fournisseur #:  37578
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Catalog # CAPI37578
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific

Blocking Buffer, 100 mL, ELISA, Western Blot, Dot Blot, Immunohistochemistry

CAS Number:  
Price
$140.26
Log in pour la disponibilité
Each (100ml)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.

  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS

StartingBlock Blocking Buffers performs well with an extremely wide range of antibodies, antibody combinations, other protein probing and assay systems.

These buffers are biotin- and serum-free. They are compatible with many detection systems and feature short blocking times. These buffers also allow stripping and reprobing with reblocking to save time and money.

Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.

Documents

Spécifications

Spécifications

  • Désignation:
    Blocking Buffer
  • Taille:
    100 mL
  • Utilisation:
    ELISA, Western Blot, Dot Blot, Immunohistochemistry

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.