Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific | Avantor
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
PBS with Tween® 20, 1 L, ELISA, Western Blot, Dot Blot, Immunohistochemistry
Catalog # CAPI37539
Supplier:   Thermo Scientific
Fournisseur #:  37539
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Catalog # CAPI37539
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
PBS with Tween® 20, 1 L, ELISA, Western Blot, Dot Blot, Immunohistochemistry
CAS Number:  
Price
$664.76
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS
StartingBlock Blocking Buffers performs well with an extremely wide range of antibodies, antibody combinations, other protein probing and assay systems.
These buffers are biotin- and serum-free. They are compatible with many detection systems and feature short blocking times. These buffers also allow stripping and reprobing with reblocking to save time and money.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.
Documents
Spécifications
Spécifications
  • Désignation:
    PBS with Tween® 20
  • Taille:
    1 L
  • Utilisation:
    ELISA, Western Blot, Dot Blot, Immunohistochemistry