Microlink® Screw Caps, WHEATON | Avantor
Microlink® Screw Caps, WHEATON
PP/Silicone/PTFE liners, 38-430, PP, White
Catalog # 89508-346
Fournisseur #:  240256-01
Microlink® Screw Caps, WHEATON
Catalog # 89508-346
Microlink® Screw Caps, WHEATON
Microlink® Screw Caps, WHEATON
PP/Silicone/PTFE liners, 38-430, PP, White
Price
$332.15
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 24
Product Details & Documents
These screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
  • Liner is ultrasonically welded to the cap to eliminate the possibility of glue contamination and bond failure common with autoclaving
  • Available in convenient lab pack quantities as well as larger case packs
  • Low-compression-set silicone ideal for the storage of samples for prolonged period of time over wide temperature ranges
  • PTFE or polypropylene on the exposed face of the liner imparts an inert,chemical-resistant surface
  • Open top caps provide access with a syringe
  • Autoclavable
Documents
Spécifications
  • Taille du bouchon:
    38-430
  • Color:
    White
  • Désignation:
    PP/Silicone/PTFE liners
  • Matière:
    PP