Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Hoffman Electrolysis Apparatus

Hoffman Electrolysis Apparatus

Hoffman Electrolysis Apparatus, 24"
  470213-364
Supplier #:  HEA001
Hoffman Electrolysis Apparatus
  470213-364
Hoffman Electrolysis Apparatus
Hoffman Electrolysis Apparatus

Hoffman Electrolysis Apparatus, 24"

 
$602.75
   
Each
 

The Hoffman Electrolysis Apparatus is designed for the electrolysis of water.

  • Apparatus Comes Equipped with All Necessary Materials
  • Designed for the Electrolysis Of Water
  • Glass Electrolysis Unit Features Two 50 mL Burets

It's also used for the qualitative and quantitative study of the electrolytic decomposition of electrolytes such as water and hydrochloric acid.

The apparatus includes all necessary materials including two 50 mL burets

Ordering information: This kit includes 1 glass electrolysis unit, 1 support stand, 1 burette clamp, 1 extension clamp, 1 bosshead clamp, 1 pair of platinum electrodes, 1 pair of carbon electrodes, and 1 activity guide.

 
  • Désignation:
    Hoffman Electrolysis Apparatus
  • Taille:
    24"
Accessories

 

  •  
    Description
     
     
  • 470345-638
    12 V DC power supply
       
    Each - $255.89

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.