Ion Selective Electrodes, Hach
Ion Selective Electrodes, Hach
sensION+ ISE Electrode, Nitrate, sensor only, no reference, 0...50 °C
Catalog # 89501-156
Product available on GSA Advantage®
Ion Selective Electrodes, Hach
Catalog # 89501-156
Supplier:  
Product available on GSA Advantage®
Ion Selective Electrodes, Hach
Ion Selective Electrodes, Hach
sensION+ ISE Electrode, Nitrate, sensor only, no reference, 0...50 °C
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
9662C NITRATE ISE COMBINATION Catalog #CA89501-156
Product CA89501-156 is a replacement for 89501-156 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Hach offers a wide range of electrodes for every application and budget
Our wide selection of metal electrodes, combined metal electrodes, and ion selective electrodes will have the right probe for your application.
Certifications: .CE.WEEE.
Ordering information: The sensION+ electrodes require a reference electrode for use. Many ISE electrodes require standards, filling solutions, ISA, etc. for electrode use.
Spécifications
  • Plage de température:
    0...50 °C
  • Désignation:
    sensION+ ISE Electrode, Nitrate, sensor only, no reference
  • Cat. No.:
    89501-156
  • Connection:
    BNC
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Chloride Standard Solutions as Cl-(NIST) 12,500 mg/L, 16/Voluette Ampules
    Chloride Standard Solutions as Cl-(NIST) 12,500 mg/L, 16/Voluette Ampules
    CA89173-990
    Log in pour la disponibilité
    Pack of 16 (10ml) - $258.43