Ion Selective Electrodes, Hach
Product CA89501-156 is a replacement for 89501-156 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Hach offers a wide range of electrodes for every application and budget
Our wide selection of metal electrodes, combined metal electrodes, and ion selective electrodes will have the right probe for your application.
Certifications: .CE.WEEE.
Ordering information: The sensION+ electrodes require a reference electrode for use. Many ISE electrodes require standards, filling solutions, ISA, etc. for electrode use.
Spécifications
- Plage de température:0...50 °C
- Désignation:sensION+ ISE Electrode, Nitrate, sensor only, no reference
- Cat. No.:89501-156
- Connection:BNC
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Chloride Standard Solutions as Cl-(NIST) 12,500 mg/L, 16/Voluette AmpulesChloride Standard Solutions as Cl-(NIST) 12,500 mg/L, 16/Voluette AmpulesCA89173-990Log in pour la disponibilitéPack of 16 (10ml) - $258.43