Variable Volume Pipettes, Hach
Variable Volume Pipette Set with Tips, 1× each 0.2 - 1.0 ml and 1.0 - 5.0 ml
Catalog # 89501-142
Jump to:
Discontinued
Product Details & Documents
Hach's variable volume single-channel, air-displacement pipettes feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation.
Outstanding accuracy with adjustments in 50 L increments (0.05 ml).
Ordering information: Supplied with calibration report and maintenance tool.
Spécifications
- Volume:1× each 0.2 - 1.0 ml and 1.0 - 5.0 ml
- Désignation:Variable Volume Pipette Set with Tips
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



