SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
These compact, easy-to-use cassettes incorporate a unique recessed cover, which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1mm mesh holes are specially designed to hold biopsy specimens, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle for use with certain types of cassette labeling instruments
- Color:Blue