Ceramic Rods, Round, Solid, AL23, Thermo Scientific
400 mm, 10 mm
  CAAA32937-QAT
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
  CAAA32937-QAT
 :  
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
400 mm, 10 mm
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
  • Ø:
    10 mm
  • Longueur:
    400 mm