Ceramic Rods, Round, Solid, AL23, Thermo Scientific
400 mm, 5 mm
  CAAA32931-QAT
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
  CAAA32931-QAT
 :  
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
400 mm, 5 mm
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
  • Ø:
    5 mm
  • Longueur:
    400 mm